Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TwdlQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143239.2 | NP_651496.1 | 2 | 739 | 70 | |
GenBank mRNA | BT100154.4 | - | 1 | 672 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |