Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038805.2 | NP_001033894.1 | 8 - 9 | 1315 | 89 | |
GenBank mRNA | AY226541.1 | - | 8 - 9 | 1120 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |