Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klhl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015100.2 | NP_001015100.2 | 787 | 105 | ||
NM_001015101.2 | NP_001015101.2 | 762 | 105 | |||
GenBank mRNA | BT014923.1 | - | 787 | 105 | ||
BT022090.2 | - | 762 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |