Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015473.3 | NP_001015473.1 | 3 - 4 | 956 | 70 | |
NM_001110929.2 | NP_001104399.1 | 3 - 4 | 956 | 70 | ||
NM_001110930.2 | NP_001104400.1 | 3 - 4 | 956 | 70 | ||
NM_001110931.2 | NP_001104401.2 | 2 - 3 | 487 | 70 | ||
GenBank mRNA | BT021347.1 | - | 3 - 4 | 772 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |