Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cht10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042957.2 | NP_001036422.1 | 8 - 9 | 5703 | 102 | |
NM_001273768.1 | NP_001260697.1 | 8 - 9 | 5787 | 102 | ||
NM_001273769.1 | NP_001260698.1 | 8 - 9 | 5774 | 102 | ||
GenBank mRNA | BT021372.1 | - | 8 - 9 | 5703 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |