Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042973.3 | NP_001036438.1 | 4 - 5 | 543 | 99 | |
NM_001169580.2 | NP_001163051.1 | 5 - 6 | 458 | 99 | ||
NM_001299206.1 | NP_001286135.1 | 4 - 5 | 543 | 99 | ||
GenBank mRNA | BT022770.1 | - | 4 - 5 | 476 | 99 | |
BT022801.1 | - | 4 - 5 | 518 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |