Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297915.1 | NP_001284844.1 | 2 - 3 | 224 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |