Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adgrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001845.2 | NP_001001845.2 | 52 - 53 | 11231 | 84 | |
XM_005165511.4 | XP_005165568.1 | 52 - 53 | 11350 | 84 | ||
XM_009301753.3 | XP_009300028.1 | 53 - 54 | 11372 | 84 | ||
GenBank mRNA | AY540131.1 | - | 52 - 53 | 11235 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |