Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adgrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001845.2 | NP_001001845.2 | 54 - 55 | 11644 | 126 | |
XM_005165511.4 | XP_005165568.1 | 54 - 55 | 11763 | 126 | ||
XM_009301753.3 | XP_009300028.1 | 55 - 56 | 11785 | 126 | ||
GenBank mRNA | AY540131.1 | - | 54 - 55 | 11648 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |