Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adgrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001845.2 | NP_001001845.2 | 70 - 71 | 14787 | 78 | |
XM_005165511.4 | XP_005165568.1 | 70 - 71 | 14906 | 78 | ||
XM_009301753.3 | XP_009300028.1 | 71 - 72 | 14928 | 78 | ||
GenBank mRNA | AY540131.1 | - | 70 - 71 | 14791 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |