Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adgrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001845.2 | NP_001001845.2 | 74 - 75 | 16072 | 74 | |
XM_005165511.4 | XP_005165568.1 | 74 - 75 | 16191 | 74 | ||
XM_009301753.3 | XP_009300028.1 | 75 - 76 | 16213 | 74 | ||
GenBank mRNA | AY540131.1 | - | 74 - 75 | 16076 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |