Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adgrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001845.2 | NP_001001845.2 | 78 - 79 | 17016 | 85 | |
XM_005165511.4 | XP_005165568.1 | 78 - 79 | 17135 | 85 | ||
XM_009301753.3 | XP_009300028.1 | 79 - 80 | 17157 | 85 | ||
GenBank mRNA | AY540131.1 | - | 78 - 79 | 85 |
Gene Symbol: LOC101886920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005174647.3 | XP_005174704.3 | 1 - 2 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |