Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adgrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001845.2 | NP_001001845.2 | 80 - 81 | 17446 | 99 | |
XM_005165511.4 | XP_005165568.1 | 80 - 81 | 17565 | 99 | ||
XM_009301753.3 | XP_009300028.1 | 81 - 82 | 17587 | 99 | ||
GenBank mRNA | AY540131.1 | - | 80 - 81 | 17450 | 99 |
Gene Symbol: LOC101886920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005174647.3 | XP_005174704.3 | 3 - 4 | 894 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |