Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adgrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001845.2 | NP_001001845.2 | 83 - 84 | 17857 | 97 | |
XM_005165511.4 | XP_005165568.1 | 83 - 84 | 17976 | 97 | ||
XM_009301753.3 | XP_009300028.1 | 84 - 85 | 17998 | 97 | ||
GenBank mRNA | AY540131.1 | - | 83 - 84 | 17861 | 97 | |
BC090691.1 | - | 4 - 5 | 447 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |