Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adgrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001845.2 | NP_001001845.2 | 44 - 45 | 9892 | 96 | |
XM_005165511.4 | XP_005165568.1 | 44 - 45 | 10011 | 96 | ||
XM_009301753.3 | XP_009300028.1 | 45 - 46 | 10033 | 96 | ||
GenBank mRNA | AY540131.1 | - | 44 - 45 | 9896 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |