Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdm5bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002166.2 | NP_001002166.2 | 15 - 16 | 2324 | 115 | |
XM_005167024.4 | XP_005167081.1 | 16 - 17 | 3002 | 115 | ||
XM_009303989.3 | XP_009302264.1 | 16 - 17 | 3002 | 115 | ||
GenBank mRNA | BC071280.1 | - | 15 - 16 | 2257 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |