Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002558.1 | NP_001002558.1 | 2 - 3 | 329 | 63 | |
GenBank mRNA | BC076244.1 | - | 2 - 3 | 329 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |