Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oclnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008618.1 | NP_001008618.1 | 8 - 9 | 1424 | 145 | |
XM_005171844.4 | XP_005171901.1 | 7 - 8 | 1521 | 145 | ||
GenBank mRNA | BC086743.1 | - | 8 - 9 | 1424 | 145 | |
BC164682.1 | - | 7 - 8 | 1278 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |