Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: supt7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013292.2 | NP_001013310.1 | 3 - 4 | 916 | 99 | |
GenBank mRNA | BC090267.1 | - | 3 - 4 | 920 | 99 | |
BC165795.1 | - | 2 - 3 | 788 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |