Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020576.2 | NP_001018412.1 | 4 - 5 | 387 | 66 | |
XM_005166692.4 | XP_005166749.1 | 3 - 4 | 364 | 66 | ||
XM_005166693.4 | XP_005166750.1 | 4 - 5 | 503 | 66 | ||
XM_005166694.4 | XP_005166751.1 | 4 - 5 | 465 | 66 | ||
XM_005166695.4 | XP_005166752.1 | 3 - 4 | 443 | 66 | ||
XM_017357334.2 | XP_017212823.1 | 3 - 4 | 696 | 66 | ||
XM_021478258.1 | XP_021333933.1 | 4 - 5 | 403 | 66 | ||
XM_021478259.1 | XP_021333934.1 | 3 - 4 | 356 | 66 | ||
GenBank mRNA | BC095208.1 | - | 4 - 5 | 391 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |