Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gxylt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030099.1 | NP_001025270.1 | 1 - 2 | 258 | 78 | |
GenBank mRNA | BC139590.1 | - | 1 - 2 | 263 | 78 | |
BC163763.1 | - | 1 - 2 | 298 | 78 | ||
BC163768.1 | - | 1 - 2 | 298 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |