Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ripply1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039108.1 | NP_001034197.1 | 2 - 3 | 230 | 85 | |
GenBank mRNA | AB212219.1 | - | 2 - 3 | 230 | 85 | |
BC162410.1 | - | 2 - 3 | 187 | 85 | ||
BC162427.1 | - | 2 - 3 | 187 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |