Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ripply1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039108.1 | NP_001034197.1 | 3 - 4 | 308 | 99 | |
GenBank mRNA | AB212219.1 | - | 3 - 4 | 308 | 99 | |
BC162410.1 | - | 3 - 4 | 265 | 99 | ||
BC162427.1 | - | 3 - 4 | 265 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |