Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nsrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077430.2 | NP_001070898.1 | 3 - 4 | 177 | 81 | |
GenBank mRNA | BC076130.1 | - | 3 - 4 | 197 | 81 | |
BC124124.1 | - | 3 - 4 | 182 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |