Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: il19l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082955.1 | NP_001076424.1 | 3 - 4 | 395 | 107 | |
GenBank mRNA | AB219579.1 | - | 3 - 4 | 395 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |