Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snrpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173251.2 | NP_775358.1 | 4 - 5 | 293 | 156 | |
GenBank mRNA | AY099526.1 | - | 4 - 5 | 295 | 156 | |
BC091442.1 | - | 4 - 5 | 293 | 156 | ||
BC165527.1 | - | 3 - 4 | 285 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |