Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hagh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200043.2 | NP_956337.1 | 3 - 4 | 319 | 75 | |
XM_005163865.4 | XP_005163922.1 | 3 - 4 | 383 | 75 | ||
XM_009299331.3 | XP_009297606.1 | 3 - 4 | 383 | 75 | ||
XM_021469713.1 | XP_021325388.1 | 3 - 4 | 383 | 75 | ||
GenBank mRNA | AY398356.1 | - | 3 - 4 | 319 | 75 | |
BC060913.1 | - | 2 - 3 | 207 | 75 | ||
BC066607.1 | - | 3 - 4 | 305 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |