Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snrnp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200322.1 | NP_956616.1 | 6 - 7 | 856 | 141 | |
GenBank mRNA | BC051783.1 | - | 6 - 7 | 856 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |