Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ilf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213236.2 | NP_998401.1 | 12 - 13 | 1010 | 110 | |
GenBank mRNA | BC059456.1 | - | 12 - 13 | 981 | 110 | |
BC066394.1 | - | 12 - 13 | 1007 | 110 | ||
EH577521.1 | - | 2 - 3 | 558 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |