Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: akr1b1.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002048.1 | NP_001002048.1 | 1 - 2 | 143 | 86 | |
GenBank mRNA | BC071313.1 | - | 1 - 2 | 143 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |