Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: quo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004109.2 | NP_001004109.2 | 8 - 9 | 2918 | 81 | |
GenBank mRNA | AY654285.2 | - | 8 - 9 | 2917 | 81 | |
BC162633.1 | - | 8 - 9 | 2838 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |