Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klhl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005316.2 | NP_001005316.2 | 5 - 6 | 1362 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Danio rerio Probe spans exons |