Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cab39l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007327.1 | NP_001007328.1 | 4 - 5 | 631 | 81 | |
XM_005170986.4 | XP_005171043.1 | 5 - 6 | 1079 | 81 | ||
GenBank mRNA | BC085674.1 | - | 4 - 5 | 631 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |