Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arhgef7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008624.1 | NP_001008624.1 | 9 - 10 | 1184 | 97 | |
XM_017354714.2 | XP_017210203.1 | 9 - 10 | 1363 | 97 | ||
XM_017354716.2 | XP_017210205.1 | 9 - 10 | 996 | 97 | ||
XM_017354718.2 | XP_017210207.1 | 9 - 10 | 959 | 97 | ||
XM_021474150.1 | XP_021329825.1 | 9 - 10 | 1069 | 97 | ||
GenBank mRNA | BC086736.1 | - | 9 - 10 | 1184 | 97 | |
BC165668.1 | - | 9 - 10 | 1103 | 97 | ||
DQ656108.1 | - | 9 - 10 | 970 | 97 | ||
DQ656109.1 | - | 9 - 10 | 1138 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |