Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: angptl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014820.1 | NP_001014820.1 | 1 - 2 | 796 | 56 | |
XM_005160416.4 | XP_005160473.1 | 2 - 3 | 1263 | 56 | ||
GenBank mRNA | AB199647.1 | - | 1 - 2 | 796 | 56 | |
BC151871.1 | - | 2 - 3 | 1181 | 56 | ||
BC163080.1 | - | 1 - 2 | 838 | 56 | ||
BC163101.1 | - | 1 - 2 | 838 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |