Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017603.1 | NP_001017603.1 | 5 - 6 | 667 | 100 | |
XM_005166840.4 | XP_005166897.1 | 5 - 6 | 789 | 100 | ||
GenBank mRNA | BC092849.1 | - | 5 - 6 | 667 | 100 | |
BC164606.1 | - | 4 - 5 | 521 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |