Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: agpat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076745.2 | NP_001070213.2 | 1 - 2 | 562 | 114 | |
GenBank mRNA | BC124532.1 | - | 1 - 2 | 357 | 114 | |
BC164409.1 | - | 1 - 2 | 248 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |