Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atad5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082829.2 | NP_001076298.2 | 3 - 4 | 1293 | 143 | |
XM_005166177.4 | XP_005166234.1 | 3 - 4 | 1358 | 143 | ||
GenBank mRNA | BC057535.1 | - | 3 - 4 | 1293 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |