Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptpn11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200454.1 | NP_956748.1 | 3 - 4 | 414 | 82 | |
XM_005162157.4 | XP_005162214.1 | 3 - 4 | 452 | 82 | ||
XM_009296835.3 | XP_009295110.1 | 3 - 4 | 606 | 82 | ||
GenBank mRNA | BC055139.1 | - | 3 - 4 | 414 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |