Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zcchc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200544.2 | NP_956838.2 | 2 - 3 | 314 | 122 | |
GenBank mRNA | AY253298.1 | - | 2 - 3 | 299 | 122 | |
BC049016.1 | - | 2 - 3 | 317 | 122 | ||
BC056336.1 | - | 2 - 3 | 315 | 122 | ||
BC068347.1 | - | 2 - 3 | 306 | 122 | ||
EH505718.1 | - | 2 - 3 | 319 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |