Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc35a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200654.1 | NP_956948.1 | 4 - 5 | 469 | 132 | |
XM_005167823.4 | XP_005167880.1 | 4 - 5 | 487 | 132 | ||
GenBank mRNA | BC057491.1 | - | 4 - 5 | 469 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |