Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cry-dash | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205686.1 | NP_991249.1 | 3 - 4 | 330 | 102 | |
GenBank mRNA | AB120759.1 | - | 3 - 4 | 330 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |