Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205754.1 | NP_991317.1 | 4 - 5 | 577 | 61 | |
GenBank mRNA | AY398401.1 | - | 4 - 5 | 577 | 61 | |
BC094958.1 | - | 4 - 5 | 564 | 61 | ||
BC165057.1 | - | 4 - 5 | 564 | 61 | ||
DQ388034.1 | - | 4 - 5 | 530 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |