Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prpf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212655.2 | NP_997820.2 | 20 - 21 | 2816 | 111 | |
GenBank mRNA | BC056710.1 | - | 20 - 21 | 2792 | 111 | |
BC066556.1 | - | 20 - 21 | 2780 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |