Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213245.1 | NP_998410.1 | 6 - 7 | 682 | 75 | |
XM_005160456.2 | XP_005160513.1 | 6 - 7 | 796 | 75 | ||
GenBank mRNA | BC063962.1 | - | 6 - 7 | 682 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |