Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctnnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001889.1 | NP_001001889.1 | 15 - 16 | 2526 | 81 | |
GenBank mRNA | AF329680.1 | - | 15 - 16 | 2526 | 81 | |
BC056276.1 | - | 15 - 16 | 2523 | 81 | ||
BC114257.1 | - | 8 - 9 | 1275 | 81 | ||
BC134896.1 | - | 7 - 8 | 877 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |