Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctnnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001889.1 | NP_001001889.1 | 5 - 6 | 898 | 59 | |
GenBank mRNA | AF329680.1 | - | 5 - 6 | 898 | 59 | |
BC056276.1 | - | 5 - 6 | 895 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |