Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rs1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003438.2 | NP_001003438.1 | 2 - 3 | 93 | 70 | |
GenBank mRNA | BC076183.1 | - | 2 - 3 | 94 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |