Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rs1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003438.2 | NP_001003438.1 | 4 - 5 | 360 | 68 | |
GenBank mRNA | BC076183.1 | - | 4 - 5 | 361 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |