Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rs1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003438.2 | NP_001003438.1 | 5 - 6 | 551 | 90 | |
GenBank mRNA | BC076183.1 | - | 5 - 6 | 552 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |